Flessen, brede hals
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1688 1689 1690 1691 1692
KART1688EA 219 EUR
KART1688 KART1689 KART1690 KART1691 KART1692
Flessen, brede hals
Flessen Kunststof flessen
PFA, translucent.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Pressure: no pressure
  • Non porous