Slangen, PVC, Isoflex
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1760 1761 1762 1763 1764 1765 1766 1767 1768
KART1760EA 35.5 EUR
KART1760 KART1761 KART1762 KART1763 KART1764 KART1765 KART1766 KART1767 KART1768
Slangen, PVC, Isoflex
Slangen Transfer Tubing
PVC, transparent.
  • Non-toxic
  • Non ageing
  • Non oxiding
The tubing is marked every meter for ease of use.
Shore 'A' durometer hardness: 75 ±2.
Leveringsinformatie: Supplied in 20 meter rolls packed in PE bags.