Voorraadflessen, rechthoekig, met kraan
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1770 1771
KART1770EA 285 EUR
KART1770 KART1771
Voorraadflessen, rechthoekig, met kraan
Flessen Voorraadflessen
Made of HDPE, translucent.
  • Comfortable handle for displacement
  • With spigot
  • Suitable for foodstuff
These two products, with size 10 liters and 20 liters, are specifically designed to rationalize laboratory and small production line organization, giving the opportunity to optimize water, aqueous solutions and solvents availability in a reduced space.