4-Nitrobenzaldehyde
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01023.1
SB01023.10
SB01023.50
MAYBSB01023.1EA
0
EUR
MAYBSB01023.1
MAYBSB01023.10
MAYBSB01023.50
4-Nitrobenzaldehyde
4-Nitrobenzaldehyde
Formule:
C₇H₅NO₃ Kookpunt: 300 °C (1013 hPa) Smeltpunt: 106 °C Dichtheid: 1,546 g/cm³ (20 °C) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007346 CAS nummer: 555-16-8 EINECS: 209-084-5 Merck Index: 12,06684 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...