Kleurbakken, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
290750002 213170003
SCOT290750002EA 33.9 EUR
SCOT290750002 SCOT213170003
Kleurbakken, DURAN®
Kleurbakken
The removable staining tray with open bottom allows quick immersion and emptying. The stainless steel handle allows the staining tray to be removed without direct contact with the staining solution.
  • The removable tray has an open bottom to facilitate rapid immersion and drainage, reducing carry-over
  • It is not necessary to take out the slides for drying
  • Allows the slides to be transported from one chamber to another
Staining tray colouring of up to 10 slides with standard dimensions 76×26 mm simultaneously or any width up to 52 mm possible.
Microscopy is a method of viewing objects that lie outside the resolution of the human eye and can be examined. The objects to be observed are magnified to such an extent that they can be displayed as images. For sample preparation, the objects to be examined are cut into very thin slices. These sections are then applied to an object slide and covered with a cover glass and appropriate solutions. To view the sections under the microscope, the sections on the slide must be filled in a staining trough and stained with an appropriate staining solution.
Stainless steel handle can be attached separately and not permanently to the dye rack, making it easy to insert and remove the slides. Allows the staining rack to be safely placed in the staining box while direct contact with the staining solution can be avoided.