Maatkolven, klasse A, wit schaalverdeling, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216787309 216780704 216782405 216780807 216781203 216781409 216781709 216782508 216783204 216784406 216785402 216786304 216783607
612-5212EA 159 EUR
612-5212 612-4998 612-5205 612-4999 612-5202 612-5203 612-5204 612-5206 612-5207 612-5209 612-5210 612-5211 612-5208
Maatkolven, klasse A, wit schaalverdeling, DURAN®
Kolven Maatkolven
DURAN®, borosilicate glass 3.3, clear, with PE stopper, without certificate of conformity.
  • With circular graduation mark
  • White printed image
  • With batch certificate
Typical applications precise measurement of specified liquid amounts, preparation and storage of standard solutions.
With printed retrace code (batch identification).
*Brede hals
Certificaten: ISO 1042