Maatcilinders, klasse B, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
213965405 213966307 213963601 213960707 213960801 213961403 213961703 213962408 213964409
215-3646EA 61 EUR
215-3646 215-3647 215-3644 215-3639 215-3640 215-3641 215-3642 215-3643 215-3645
Maatcilinders, klasse B, DURAN®
Cilinders Maatcilinders
DURAN®, borosilicaatglas 3.3, helder., klasse B.
  • Calibrate to contain ('In')
  • Marks and inscription in white
  • With spout
  • With large hexagonal base
Base ist equipped with three knobs to increase the stability
Certificaten: DIN ISO 4788