Maatcilinders, laag model, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
213950809 213951402 213951702 213955404 213956306 213952407 213953609 213954408
215-3631EA 25.1 EUR
215-3631 215-3632 215-3633 215-3637 215-3638 215-3634 215-3635 215-3636
Maatcilinders, laag model, DURAN®
Cilinders Maatcilinders
DURAN®, borosilicate glass 3.3, clear, class B, low form.
  • Calibrate to contain ('In')
  • Marks and inscription in white
  • With spout
  • With large hexagonal base
Base ist equipped with three knobs to increase the stability