Flessen, brede hals, met manipulatieveilige schroefdop
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15710 15711 15712
KART15710EA 3.4 EUR
KART15710 KART15711 KART15712
Flessen, brede hals, met manipulatieveilige schroefdop
Flessen Kunststof flessen
HDPE, translucent, with black screw caps.
  • Good chemical resistance
  • Good temperature resistance
The tamper-evident cap incorporates a safety ring which is secured when the cap is screwed on to the bottle. Opening the bottle breaks the safety ring, showing signs of interference.