Worktables, motorised, Concept
Leverancier: TRESTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TT12075-ESD TT10060-HPL TT10060-ESD 10349015P 10349016P 10349017P TT15060-ESD TT15060-HPL 10349018P 10349019P 10349033P 10349062P 10349063P 10349064P 10349065P 10349066P 10349030P TT10075-ESD TT10075-HPL TT10090-ESD TT10090-HPL TT12060-ESD TT12060-HPL TT12090-ESD TT12090-HPL TT15075-ESD TT15075-HPL TT18060-ESD TT18060-HPL TT18075-ESD TT18075-HPL TT18090-ESD TT18090-HPL TT20060-ESD TT20060-HPL TT20075-ESD TT20090-ESD TT20090-HPL TT20075-LPL
SOVETT12075-ESDEA 0 EUR
SOVETT12075-ESD SOVETT10060-HPL SOVETT10060-ESD SOVE10349015P SOVE10349016P SOVE10349017P SOVETT15060-ESD SOVETT15060-HPL SOVE10349018P SOVE10349019P SOVE10349033P SOVE10349062P SOVE10349063P SOVE10349064P SOVE10349065P SOVE10349066P SOVE10349030P SOVETT10075-ESD SOVETT10075-HPL SOVETT10090-ESD SOVETT10090-HPL SOVETT12060-ESD SOVETT12060-HPL SOVETT12090-ESD SOVETT12090-HPL SOVETT15075-ESD SOVETT15075-HPL SOVETT18060-ESD SOVETT18060-HPL SOVETT18075-ESD SOVETT18075-HPL SOVETT18090-ESD SOVETT18090-HPL SOVETT20060-ESD SOVETT20060-HPL SOVETT20075-ESD SOVETT20090-ESD SOVETT20090-HPL SOVETT20075-LPL
Worktables, motorised, Concept
Meubilair Tafels
The motorised sit-stand worktable is designed to meet the ergonomic requirements of today's labs; it adapts to the work process and the needs of the user.
  • The working position can be changed from 700 to 1100 mm
  • Loading capacity is max. 400 kg
  • Control panel includes up, down
  • 3 memory buttons for storing pre-selected heights
  • Frame, tabletop, and accessories available with ESD protection
  • Colour - Light grey
The sit-stand worktable can be used as a flat workspace or in combination with heavy equipment such as laminar flow cabinets. Even suitable for clean room ISO class 7.
Bestelinformatie: Bench top needs to be ordered separately. A wide range of ergonomic accessories is available, such as adjustable shelves, LCD arms and keyboard holders. For details, please contact your local Avantor sales office.