1-kanaalspipetten, vast volume, Fixopet
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1303.10 1304.10 1305.10 1306.10 1307.10 1308.10 1309.10 1310.10 1311.10 1312.10 1313.10
KART1303.10EA 316 EUR
KART1303.10 KART1304.10 KART1305.10 KART1306.10 KART1307.10 KART1308.10 KART1309.10 KART1310.10 KART1311.10 KART1312.10 KART1313.10
1-kanaalspipetten, vast volume, Fixopet
Pipetten Micropipetten
These single channel pipettes are available with fixed volumes in a range from 5 to 1000 µl.
  • Special handle ensures a more comfortable grip
  • Rotary device for the tip ejection does not create strain on the tendons of the thumb
  • Removable ejector: Can be used with test tubes with a very narrow opening
  • Integrated recalibration system ensures quick and precise recalibrations
  • Robust structure of the PVDF body ensures high mechanical and chemical resistances
  • Resistant and durable
  • Completely autoclavable at +121 °C for 20 minutes