Anthranilonitrile 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
104920100
104921000
ACRO104920100EA
24.1
EUR
ACRO104920100
ACRO104921000
Anthranilonitrile 98%
Anthranilonitrile
Formule:
C₇H₆N₂ Kookpunt: 267…268 °C (760 mmHg) Smeltpunt: 46…50 °C Dichtheid: 1,26 g/cm³ (20 °C) Vlampunt: 145 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007631 CAS nummer: 1885-29-6 EINECS: 217-549-9 UN: 3439 ADR: 6.1,III |
Specification Test Results
Appearance | Off-white to yellow to beige to brown Powder or crystals or flakes and/or chunks |
Infrared spectrum | Conforms |
Melting point | 46 to 51 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...