Pipetten met schaalverdeling, klasse B, totale afgifte, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
243442901 243440103 243440309 243440609 243441108 243442301 243443409 243441605
612-5264EA 92.9 EUR
612-5264 612-5258 612-5259 612-5260 612-5261 612-5263 612-5265 612-5262
Pipetten met schaalverdeling, klasse B, totale afgifte, DURAN®
Pipetten Gegradueerde pipetten
Soda-lime glass (AR-Glas®).
  • For cotton plug
  • Marks and inscription in brown
  • Numbering: Zero at top
  • Graduated to tip (total delivery, type 3)
  • Calibration is based on the poured out volume (Ex)
Complies to DIN ISO 835.
Colour code complies to DIN 12621.