Volpipetten, klasse AS, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
243380109 243380203 243380709 243380803 243381208 243381405 243381705 243382401
612-5242EA 9.4 EUR
612-5242 612-5243 612-5244 612-5245 612-5246 612-5247 612-5248 612-5249
Volpipetten, klasse AS, DURAN®
Pipetten Volpipetten
Natronkalkglas (AR-Glas®)
  • Marks and inscription in brown
  • Calibration is based on the poured out volume (Ex)
Complies to DIN ISO 648.
Colour code complies to DIN 12621.
Printed Retrace Code (batch identification).
*no bulb
Order Now
Specificaties
Environmentally Preferable