Quartz microfibre filters
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FT-3-1109-047 FT-3-1109-037 FT-3-1109-050 FT-3-1109-090 FT-3-1109-110 FT-3-1109-125 FT-3-1109-150 FT-3-1109-025 FT-3-1109-085
SARTFT-3-1109-047EA 130 EUR
SARTFT-3-1109-047 SARTFT-3-1109-037 SARTFT-3-1109-050 SARTFT-3-1109-090 SARTFT-3-1109-110 SARTFT-3-1109-125 SARTFT-3-1109-150 SARTFT-3-1109-025 SARTFT-3-1109-085
Quartz microfibre filters
Filters Glasvezelfilters
Quartz microfiber filters are free of glass fibers and binding agents.
  • Grade T 293, quartz microfiber filters unconditioned
  • Grade MK 360, quartz microfiber filters conditioned (heat pre-treated)
They are especially suited for emission monitoring at temperature of up to 900 °C and wherever filters of the highest purity are needed (such as for air monitoring according to US EPA PM 10 or trace element analyses).
Certificaten: Tested and classified according to the standards EN 1822-1:2009