Sleeves for filter crucibles, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
292013109 292011408 292012104 292012601
201-0158EA 31.3 EUR
201-0158 201-0156 SCOT292012104 201-0157
Sleeves for filter crucibles, DURAN®
Steunen Steunhoezen
These sleeves are made from EPDM (ethyene/propylene-diene-rubber).