Gaswasflessen, met kop, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
257010107 257020108
SCOT257010107EA 231 EUR
SCOT257010107 SCOT257020108
Gaswasflessen, met kop, DURAN®
Flessen Gaswasflessen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • With fused-in filter disc
  • With standard ground joint
Gas washing bottles, head with fritted disc and standard ground joint.
Order Now
Specificaties
Environmentally Preferable