Admix-naalden, BD Nokor™
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
300780 300779
BDAM300780EA 76.9 EUR
BDAM300780 BDAM300779
Admix-naalden, BD Nokor™
Injectienaalden
The BD Nokor™ Admix Needles are single-use medical devices used for the aspiration of fluids from vials and ampoules. These needles are not intended for skin injections.
  • Vented point design features a cannula side port
  • Design allows fluid placement on the container sidewall during reconstitution, helping reduce foaming or spray
  • Sterile, single-use design
The BD Nokor™ Admix needles has a cannula side port to place fluid on the sidewall of a container during reconstitution that helps reduce foaming or spray of powdered medication.