ORP standards for Redox/ORP electrodes, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
967961 967901
ORIO967961EA 195 EUR
ORIO967961 ORIO967901
ORP standards for Redox/ORP electrodes, Orion™
Standaarden Redox- en ORP-standaarden
Measure ORP simply and accurately with Thermo Scientific™ Orion™ ORP standards for Redox/ORP electrodes
  • Does not contain hazardous chemicals
  • Included in the new ASTM ORP method
  • Used as relative mV standard to calibrate the offset of sample
Orion™ OPR standards are designed for use with ORP/ATC Triode™ electrodes and Sure-Flow™ ORP electrodes.
Stable for up to a year if unopened and can be used directly from the bottle with no mixing or diluting.