Statiefkoppelingen, globulair
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2360 2362 2363
USBE2360EA 11.9 EUR
USBE2360 USBE2362 USBE2363
Statiefkoppelingen, globulair
Klemmen Statiefkoppelingen
Bossheads made of zinc die-cast, either natural, powder coated or nickel plated.
  • 35 mm Ø
  • For rods up to 13 mm Ø
  • Nickel plated hinged screws
Weight: 100 g