Micro filtertrechters, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
258575104 258575207 258575301 258575404
SCOT258575104EA 37.6 EUR
SCOT258575104 SCOT258575207 SCOT258575301 SCOT258575404
Micro filtertrechters, DURAN®
Trechters Filtertrechters Microfiltertrechters
DURAN®, borosilicate glass 3.3, clear.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
With integrated sintered glass filter plate.