Filterbuizen volgens Allihn, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
258532103 258532206 258532309 258532403
SCOT258532103EA 401 EUR
SCOT258532103 SCOT258532206 SCOT258532309 SCOT258532403
Filterbuizen volgens Allihn, DURAN®
Trechters Filtertrechters Microfiltertrechters
DURAN®, borosilicate glass 3.3, clear.
  • For filtration of very small volumes
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Very good temperature resistance
With integrated sintered glass filter plate.