Orale spuiten, BD™
Leverancier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
305207 305209 305210 305217 305219 305220
BDAM305207EA 293 EUR
BDAM305207 BDAM305209 BDAM305210 BDAM305217 BDAM305219 BDAM305220
Orale spuiten, BD™
Spuiten Injectiespuiten, medisch
BD® oral/enteral syringe with BD UniVia™ connection is intended for use by healthcare professionals to measure and administer oral medication and enteral nutrition.
  • Non-luer compatible BD UniVia™ connection designed to reduce the risk of tubing set misconnections
  • Amber coloured syringes protect light-sensitive drugs
  • Oral syringes are used for the transport and administration of non-injectable liquid medication
  • Can be used in paediatric departments
  • Natural rubber latex, BPS, or DEHP are not part of the material formulation
  • Non-sterile
The plunger rod and labeling are colored orange to aid in distinguishing the BD Oral/Enteral syringe from syringes with a luer (6%) taper.