Petrischalen, vierkant, Corning® Gosselin™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BP124-05 BP124-04
GOSSBP124-05EA 354 EUR
GOSSBP124-05 391-0444
Petrischalen, vierkant, Corning® Gosselin™
Cultuurschalen Petrischalen
PS. Petrischalen gemaakt onder aseptische cleanroomomstandigheden.
  • Manufactured from optically clear virgin polystyrene
  • Sterilised by irradiation to SAL 10⁻³
  • Supplied with 4 vents to provide consistent gas exchange
  • New design to enhance stacked dish stability
Corning® Gosselin™ square petri dishes are optically clear, with a design that has been optimised for use on automated platforms.
The square design is used in colony picking and antibiogram applications.