Kolven, platte bodem, met genormaliseerde slijpingen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241714408 241716306 241712407 241712604 241712707 241713609 241713703 241714605 241715404 241715601
SCOT241714408EA 300 EUR
SCOT241714408 SCOT241716306 SCOT241712407 SCOT241712604 SCOT241712707 SCOT241713609 SCOT241713703 SCOT241714605 SCOT241715404 SCOT241715601
Kolven, platte bodem, met genormaliseerde slijpingen, DURAN®
Kolven Platbodemkolven
DURAN®, borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
*non-DIN ISO size
Certificaten: According to DIN ISO 4797
Order Now
Specificaties
Environmentally Preferable