Veren voor aluminium ringen
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
01102825 01102835 01102840 01102860
LENZ01102825EA 2.7 EUR
LENZ01102825 801-1731 LENZ01102840 LENZ01102860
Veren voor aluminium ringen
Veren
Stainless steel springs with two rings for the safe connection of conical ground joints in combination with alu rings with hooks.