Kel-F® hub needles
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
90118 90121 90122 90123 90124 90125 90126 90138 90139 90518 90522 90526 90134 90539 7750-19 7752-09 90524 7746-10 7750-18 7746-11 90534 90120 7750-09 7750-11 7750-13 7746-07 7750-12 90521 90533 7750-04 90517 7746-06 90523 90516 90520 90528 90525 7746-02 7746-01 7746-03 7746-04 7746-05 7746-08 7746-09 7746-12 7750-23 7750-05 7750-02 7750-22 7750-01 7750-08 7750-15 7750-17 7750-21 7752-01 7752-02 7752-03 7752-04 7752-07 7752-05 7752-08 7752-11 7752-12 7752-13 7752-14 7752-15 7752-16 7752-17 7752-18 7752-19 7750-03 7750-07 7750-14 7750-16 7750-24 7750-20 90514
HAMI90118EA 127 EUR
HAMI90118 HAMI90121 613-0102 HAMI90123 HAMI90124 HAMI90125 613-0115 613-0101 HAMI90139 HAMI90518 HAMI90522 HAMI90526 HAMI90134 HAMI90539 HAMI7750-19 HAMI7752-09 HAMI90524 HAMI7746-10 HAMI7750-18 HAMI7746-11 HAMI90534 HAMI90120 HAMI7750-09 HAMI7750-11 HAMI7750-13 HAMI7746-07 HAMI7750-12 HAMI90521 HAMI90533 HAMI7750-04 HAMI90517 HAMI7746-06 HAMI90523 HAMI90516 HAMI90520 HAMI90528 HAMI90525 HAMI7746-02 HAMI7746-01 HAMI7746-03 HAMI7746-04 HAMI7746-05 HAMI7746-08 HAMI7746-09 HAMI7746-12 HAMI7750-23 HAMI7750-05 HAMI7750-02 HAMI7750-22 HAMI7750-01 HAMI7750-08 HAMI7750-15 HAMI7750-17 HAMI7750-21 HAMI7752-01 HAMI7752-02 HAMI7752-03 HAMI7752-04 HAMI7752-07 HAMI7752-05 HAMI7752-08 HAMI7752-11 HAMI7752-12 HAMI7752-13 HAMI7752-14 HAMI7752-15 HAMI7752-16 HAMI7752-17 HAMI7752-18 HAMI7752-19 HAMI7750-03 HAMI7750-07 HAMI7750-14 HAMI7750-16 HAMI7750-24 HAMI7750-20 HAMI90514
Kel-F® hub needles
Injectienaalden
Kel-F® hub (KF) needles can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Custom lengths and point styles are available.