Kolven, ronde bodem, met drie halzen en genormaliseerde slijpingen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241853602 241854401 241854504 241854607 241855509 241855603 241856308
SCOT241853602EA 72 EUR
SCOT241853602 SCOT241854401 SCOT241854504 SCOT241854607 SCOT241855509 SCOT241855603 SCOT241856308
Kolven, ronde bodem, met drie halzen en genormaliseerde slijpingen, DURAN®
Kolven Rondbodemkolven
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
One centre neck and two parallel side necks.
DIN 12392
Certificaten: DIN 12392