Ingebouwde filters
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
258550104 258550207 258552105 258552208 258550301 258550404 258551109 258551203 258551306 258551409 258552302
511-0401EA 65.9 EUR
511-0401 511-0402 511-0421 511-0422 511-0403 511-0404 511-0411 511-0412 511-0413 511-0414 511-0423
Ingebouwde filters
Filters Lijnfilters
These in-line filters are used for in-line filtration of gas lines to remove solid impurities (e.g. dust).
Made from DURAN®, borosilicate glass 3,3, clear.