Condensors, spiraal, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
242537105 242537208
SCOT242537105EA 117 EUR
SCOT242537105 SCOT242537208
Condensors, spiraal, DURAN®
Condensors
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For cooling condensate from vapours, with cone and socket. Water supplied through plastic connectors.