Instrument trays
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4743 4744
USBE4743EA 58.4 EUR
USBE4743 USBE4744
Instrument trays
Reservoirs/Schalen/Potten Instrumentenschalen
Made of stainless steel.
  • Rounded corners
  • With rim
  • Very good chemical and temperature resistance