Evaporating dishes, flat form
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5012 5013
USBE5012EA 31 EUR
USBE5012 USBE5013
Evaporating dishes, flat form
Reservoirs/Schalen/Potten Uitdampschalen
Dishes made of stainless steel.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean