Venapunctiesets, Surshield™ Surflo®
Leverancier: TERUMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SV-S21NL30 SV-S19NL30 SV-S23NL30
TERUSV-S21NL30EA 85.5 EUR
TERUSV-S21NL30 TERUSV-S19NL30 TERUSV-S23NL30
Venapunctiesets, Surshield™ Surflo®
Canules
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.