Vaten, rond
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3482-0110 3482-0050 3482-0075 3482-0040
216-5625EA 213 EUR
216-5625 216-5627 BURK3482-0075 BURK3482-0040
Vaten, rond
Vaten, vaten, trommels
Tubs, round, made of HDPE. For every purpose, with strong edge and practical carrying handles for safe transport. A lid can be delivered for every tub.
  • Temperature resistant from −30 to 70 °C
  • Suitable for food
  • High degree of chemical resistance