Druppeltrechters, cilindrisch, zonder drukbalans, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
241221704 241222409 241223602 241224401 241225406
SCOT241221704EA 108 EUR
SCOT241221704 SCOT241222409 SCOT241223602 SCOT241224401 SCOT241225406
Druppeltrechters, cilindrisch, zonder drukbalans, DURAN®
Trechters Druppeltrechters
DURAN®, borosilicate glass 3.3, clear, with glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible divisions for approximate volume reading.
DIN ISO 4800