GC septa, high temperature septa
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75801 75802 75803 75804 75805 75806 75807 75813 75810
HAMI75801EA 31.4 EUR
HAMI75801 HAMI75802 HAMI75803 HAMI75804 HAMI75805 HAMI75806 HAMI75807 HAMI75813 HAMI75810
GC septa, high temperature septa
Tussenschoten
These preconditioned PTFE/silicone rubber septa are 3,175 mm thick.
  • Average life up to 100 injections with a 26s gauge needle
  • Maximum operating temperature 212 °C