Instrument trays
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4230-3024 4230-3027 4230-3036 4230-3043 4230-3019
BURK4230-3024EA 10.9 EUR
BURK4230-3024 BURK4230-3027 BURK4230-3036 BURK4230-3043 BURK4230-3019
Instrument trays
Reservoirs/Schalen/Potten Instrumentenschalen
Melamine, white.
  • Tray without lid
  • Autoclavable
  • Suitable for use with foodstuffs
Bestelinformatie: Lid available as accessory