GC septa, long life GC autosampler septa
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75826 75827 75829 75830 75831 75832
HAMI75826EA 120 EUR
HAMI75826 HAMI75827 HAMI75829 HAMI75830 HAMI75831 HAMI75832
GC septa, long life GC autosampler septa
Tussenschoten
Long Life GC autosampler septa are designed for use with GC autosamplers.
  • Inlet temperature of up to 400 °C
  • Injection life of 400+ injections can be achieved with proper care
The 5, 9, 11 and 11,5 mm diameter septa are available with a unique prepierced center depression which guides the syringe needle to the same injection point. The 9,5, 12,7 and 17 mm diameter septa are supplied without the unique center depression.