Automatic burettes, Pellet pattern, lateral stopcock
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
341016060 341017060 341017560 341036060 341037060 341037560
612-1514EA 225 EUR
612-1514 612-9804 HIRS341017560 HIRS341036060 HIRS341037060 HIRS341037560
Automatic burettes, Pellet pattern, lateral stopcock
Buretten
Automatic burettes, Pellet pattern, class B, DURAN® with blue graduation.
  • Schellbach stripes for enhanced legibility
  • Lateral glass stopcock or PTFE valve stopcock
Certificaten: Caliberated for DIN EN ISO 385.