Burettes, Pellet pattern, brown, lateral stopcock
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
342016060 342017060 342017560 342036060 342037060 342037560
HIRS342016060EA 238 EUR
HIRS342016060 HIRS342017060 HIRS342017560 HIRS342036060 HIRS342037060 HIRS342037560
Burettes, Pellet pattern, brown, lateral stopcock
Buretten
Automatic burettes, Pellet pattern, class B, DURAN® brown glass, with white graduation.
  • Schellbach stripes for enhanced legibility
  • Lateral stopcock with glass key or PTFE spindle
Certificaten: Compliance with DIN EN ISO 385.