Trimellitic acid

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01864.1 SB01864.10 SB01864.50
MAYBSB01864.1EA 0 EUR
MAYBSB01864.1 MAYBSB01864.10 MAYBSB01864.50
Trimellitic acid
Trimellitic acid
Formule: C₉H₆O₆
Kookpunt: 107…113 °C (0,1 mmHg)
Smeltpunt: 240…245 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002470
CAS nummer: 528-44-9
EINECS: 208-432-3
Merck Index: 12,09832

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR