tert-Butyl perbenzoate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: tert-Butyl peroxybenzoate, Luperox(R) P, tert-Butyl peroxybenzoate

170142500 170140010
170142500.EA 44.8 EUR
170142500. 170140010.
tert-Butyl perbenzoate 98%
tert-Butyl perbenzoate
Formule: C₁₁H₁₄O₃
MW: 194.23 g/mol
Kookpunt: 75…76 °C (.2 mmHg)
Smeltpunt: 8
Dichtheid: 1.04 g/cm³
Vlampunt: 93 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008802
CAS nummer: 614-45-9
EINECS: 210-382-2
UN: 3103
ADR: 5.2

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5000 (20°C, 589 nm)
Additional info saponifiable chlorine: 0.035%
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR