tert-Butyl perbenzoate ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
tert-Butyl peroxybenzoate, Luperox(R) P, tert-Butyl peroxybenzoate
A17373.14
A17373.22
A17373.36
A17373.14EA
23.2
EUR
A17373.14
A17373.22
A17373.36
tert-Butyl perbenzoate ≥98%
tert-Butyl perbenzoate
Purity: 98%
CAS No.: 614-45-9
Molecular Formula: C11H14O3
Formula Weight: 194.23
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.4990
Odour: Sweet
Melting Point: 6-8°
Boiling Point: 75-76°/0.2mm
Beilstein Reference: 1342734
UN No.: 3103
MDL No.: MFCD00008802
Formule:
C₁₁H₁₄O₃ MW: 194.23 g/mol Kookpunt: 75…76 °C (0.2 mmHg) Smeltpunt: 6…8 °C Dichtheid: 1.042 Vlampunt: 93 °C (199 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00008802 CAS nummer: 614-45-9 EINECS: 210-382-2 UN: 3103 ADR: 5.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...