Chloorbenzeen ≥99.5% for HPLC
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Phenylchloride, Benzene Chloride, Monochlorobenzene
22922.K2
22922.M1
22922.K2EA
86.2
EUR
22922.K2
22922.M1
Chloorbenzeen ≥99.5% for HPLC
Chloorbenzeen
Purity: 99.5%
CAS No.: 108-90-7
Molecular Formula: C6H5Cl
Formula Weight: 112.56
Flash Point: 28°(82°F)
Physical Form: Liquid
Density: 1.106
Refractive Index: 1.5240
Appearance: Colourless
Odour: Aromatic
Melting Point: -45°
Boiling Point: 132°
Beilstein Reference: 605632
Merck Reference: 14,2121
UN No.: 1134
MDL No.: MFCD00000530
Formule:
C₆H₅Cl MW: 112.56 g/mol Kookpunt: 132 °C Smeltpunt: –45 °C Dichtheid: 1.106 Vlampunt: 28 °C (82 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00000530 CAS nummer: 108-90-7 EINECS: 203-628-5 UN: 1134 ADR: 3 III Bereik: 01-2119432722-45 Merck Index: 14,2121 |