2-Chloorbenzaldehyde 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
154610010 154610050 154611000 154612500 154615000
ACRO154610010EA 143 EUR
ACRO154610010 ACRO154610050 ACRO154611000 ACRO154612500 ACRO154615000
2-Chloorbenzaldehyde 99%
2-Chloorbenzaldehyde
Formule: C₇H₅ClO
MW: 140.57 g/mol
Kookpunt: 211.9
Smeltpunt: 10…11.5 °C
Dichtheid: 1.25 g/cm³
Vlampunt: 87 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003304
CAS nummer: 89-98-5
EINECS: 201-956-3
UN: 3265
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5670 (20°C, 589 nm)
GC ≥98.5 %
Free acid ≤1 % (as chlorobenzoic acid)