5-Chlorosalicylaldehyde 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 5-Chloro-2-hydroxybenzaldehyde
405030250 405031000 405030050
ACRO405030250EA 148 EUR
ACRO405030250 ACRO405031000 ACRO405030050
5-Chlorosalicylaldehyde 98%
5-Chlorosalicylaldehyde
Formule: C₇H₅ClO₂
MW: 156.57 g/mol
Kookpunt: 105 °C (12 mmHg)
Smeltpunt: 102…103 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003331
CAS nummer: 635-93-8
EINECS: 211-244-4
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 103 °C
GC ≥97.5 %
Residue after evaporation ≤0.0005 %
Free amine ≤0.0002 %