5-Chlorosalicylaldehyde
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
5-Chloro-2-hydroxybenzaldehyde
BTB02689.1
BTB02689.10
BTB02689.50
MAYBBTB02689.1EA
0
EUR
MAYBBTB02689.1
MAYBBTB02689.10
MAYBBTB02689.50
5-Chlorosalicylaldehyde
5-Chlorosalicylaldehyde
Formule:
C₇H₅ClO₂ MW: 156.57 g/mol Kookpunt: 105 °C (12 mmHg) Smeltpunt: 100.3 |
MDL Nummer:
MFCD00003331 CAS nummer: 635-93-8 EINECS: 211-244-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...