3-Chloorbenzonitril 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: m-Chlorobenzonitrile
108680100 108680500
ACRO108680100EA 31.5 EUR
ACRO108680100 ACRO108680500
3-Chloorbenzonitril 99%
3-Chloorbenzonitril
Formule: C₇H₄ClN
MW: 137.57 g/mol
Kookpunt: 94 °C (11 mmHg)
Smeltpunt: 38…42 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001798
CAS nummer: 766-84-7
EINECS: 212-172-6
Order Now
Specification Test Results
Appearance Off-white to light beige Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 38 to 42 °C
GC ≥98.5 %
Stabilizer 50 to 150 ppm Amylene