4-Chloorbenzonitril 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: p-Chlorobenzonitrile
151871000 151870250 151875000
ACRO151871000EA 75.3 EUR
ACRO151871000 ACRO151870250 ACRO151875000
4-Chloorbenzonitril 99%
4-Chloorbenzonitril
Formule: C₇H₄ClN
MW: 137.57 g/mol
Kookpunt: 223
Smeltpunt: 91…94 °C
Vlampunt: 108 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001813
CAS nummer: 623-03-0
EINECS: 210-765-4
Order Now
Specification Test Results
Appearance White to yellow Flakes and/or chunks
Infrared spectrum Conforms
Melting point 91 to 94 °C
Water ≤0.1 %
GC ≥98.5 %