2-Chloro-5-nitrobenzonitrile
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB00936.1
SB00936.10
SB00936.50
MAYBSB00936.1EA
0
EUR
MAYBSB00936.1
MAYBSB00936.10
MAYBSB00936.50
2-Chloro-5-nitrobenzonitrile
2-Chloro-5-nitrobenzonitrile
Formule:
C₇H₃ClN₂O₂ Kookpunt: 119…122 °C (0,6 mmHg) Smeltpunt: 108 °C |
MDL Nummer:
MFCD00007292 CAS nummer: 16588-02-6 EINECS: 240-643-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...